Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024535.1 | NP_001019706.1 | 1 - 2 | 437 | 136 | |
GenBank mRNA | BC119852.1 | - | 1 - 2 | 123 | 136 | |
BT021716.1 | - | 1 - 2 | 437 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC119852.1, NM_001024535.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
