Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024535.1 | NP_001019706.1 | 2 - 3 | 549 | 89 | |
GenBank mRNA | BC119852.1 | - | 2 - 3 | 235 | 89 | |
BT021716.1 | - | 2 - 3 | 549 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
