Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024535.1 | NP_001019706.1 | 4 - 5 | 820 | 70 | |
GenBank mRNA | BC119852.1 | - | 4 - 5 | 506 | 70 | |
BT021716.1 | - | 4 - 5 | 820 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
