Assay Details
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024541.2 | NP_001019712.1 | 3 - 4 | 349 | 70 | |
GenBank mRNA | AY911317.1 | - | 3 - 4 | 350 | 70 | |
BC102068.1 | - | 3 - 4 | 390 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
