Assay Details
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024558.1 | NP_001019729.1 | 1 - 2 | 410 | 62 | |
GenBank mRNA | BC150021.1 | - | 1 - 2 | 428 | 62 | |
BT021145.1 | - | 1 - 2 | 410 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
