Assay Details
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024568.2 | NP_001019739.1 | 1 - 2 | 69 | 63 | |
| GenBank mRNA | AY911366.1 | - | 1 - 2 | 70 | 63 | |
| BC102249.1 | - | 1 - 2 | 36 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102249.1, NM_001024568.2 transcripts.
Set Membership: |
|

RPS11
Genomic Map