Assay Details
| Gene Symbol: RPS11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024568.2 | NP_001019739.1 | 4 - 5 | 409 | 57 | |
| GenBank mRNA | AY911366.1 | - | 4 - 5 | 410 | 57 | |
| BC102249.1 | - | 4 - 5 | 376 | 57 | ||
| DQ347611.1 | - | 1 - 2 | 121 | 57 | ||
More Information
Set Membership: |
|

RPS11
Genomic Map