Assay Details
| Gene Symbol: PLPP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024573.1 | NP_001019744.1 | 1 | 948 | 60 | |
| GenBank mRNA | BC147999.1 | - | 1 | 997 | 60 | |
| BT021612.1 | - | 1 | 948 | 60 | ||
More Information
Set Membership: |
|

PLPP6
Genomic Map