Assay Details
Gene Symbol: NFIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024574.1 | NP_001019745.1 | 1 - 2 | 33 | 51 | |
GenBank mRNA | BC114080.1 | - | 1 - 2 | 106 | 51 | |
DQ004950.1 | - | 1 - 2 | 33 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
