Assay Details
| Gene Symbol: ECHS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025206.2 | NP_001020377.2 | 6 - 7 | 764 | 63 | |
| GenBank mRNA | AY049961.1 | - | 5 - 6 | 661 | 63 | |
| BC109605.1 | - | 6 - 7 | 759 | 63 | ||
| BT021569.1 | - | 6 - 7 | 764 | 63 | ||
| DQ058610.1 | - | 6 - 7 | 739 | 63 | ||
More Information
Set Membership: |
|

ECHS1
Genomic Map