Assay Details
| Gene Symbol: RPS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025317.1 | NP_001020488.1 | 1 - 2 | 114 | 90 | |
| GenBank mRNA | AB098931.1 | - | 1 - 2 | 117 | 90 | |
| AY911334.1 | - | 2 - 3 | 131 | 90 | ||
| BC102608.1 | - | 2 - 3 | 129 | 90 | ||
| BC126849.1 | - | 2 - 3 | 131 | 90 | ||
| BT021062.1 | - | 1 - 2 | 114 | 90 | ||
More Information
Set Membership: |
|

RPS8
Genomic Map