Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025325.2 | NP_001020496.1 | 1 - 2 | 36 | 63 | |
| GenBank mRNA | AY911369.1 | - | 1 - 2 | 37 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025325.2 transcripts.
Set Membership: |
|

PLAC8A
Genomic Map