Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025325.2 | NP_001020496.1 | 3 - 4 | 258 | 118 | |
| GenBank mRNA | AY911369.1 | - | 3 - 4 | 259 | 118 | |
| BC114195.1 | - | 2 - 3 | 174 | 118 | ||
More Information
Set Membership: |
|

PLAC8A
Genomic Map