Assay Details
| Gene Symbol: PLAC8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025325.2 | NP_001020496.1 | 5 - 6 | 497 | 134 | |
| GenBank mRNA | AY911369.1 | - | 5 - 6 | 498 | 134 | |
| BC114195.1 | - | 4 - 5 | 134 | |||
More Information
Set Membership: |
|

PLAC8A
Genomic Map