Assay Details
Gene Symbol: GMNN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025337.2 | NP_001020508.1 | 3 - 4 | 263 | 73 | |
GenBank mRNA | AY911337.1 | - | 3 - 4 | 265 | 73 | |
BC109880.1 | - | 3 - 4 | 299 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
