Assay Details
Gene Symbol: FES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032300.1 | NP_001027471.1 | 14 - 15 | 1874 | 57 | |
GenBank mRNA | BT020947.1 | - | 14 - 15 | 1874 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
