Assay Details
| Gene Symbol: RPL18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033619.2 | NP_001028791.1 | 2 - 3 | 236 | 116 | |
| GenBank mRNA | AB098916.1 | - | 2 - 3 | 211 | 116 | |
| BC102624.1 | - | 2 - 3 | 237 | 116 | ||
More Information
Set Membership: |
|

RPL18A
Genomic Map