Assay Details
| Gene Symbol: RPS3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034038.1 | NP_001029210.1 | 3 - 4 | 378 | 63 | |
| GenBank mRNA | AB098918.1 | - | 2 - 3 | 243 | 63 | |
| AY911372.1 | - | 3 - 4 | 378 | 63 | ||
| BC102789.1 | - | 1 | 374 | 63 | ||
| BC151408.1 | - | 1 | 378 | 63 | ||
More Information
Set Membership: |
|

RPS3A
Genomic Map