Assay Details
| Gene Symbol: TUBA1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034204.1 | NP_001029376.1 | 1 | 100 | 56 | |
| GenBank mRNA | BC102116.1 | - | 1 | 100 | 56 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102116.1, NM_001034204.1 transcripts.
Set Membership: |
|

TUBA1C
Genomic Map