Assay Details
| Gene Symbol: MOB3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034229.1 | NP_001029401.1 | 2 - 3 | 600 | 72 | |
| GenBank mRNA | BC123409.1 | - | 2 - 3 | 649 | 72 | |
| BT021734.1 | - | 2 - 3 | 600 | 72 | ||
More Information
Set Membership: |
|

MOB3A
Genomic Map