Assay Details
Gene Symbol: DUSP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034259.1 | NP_001029431.1 | 1 - 2 | 328 | 74 | |
GenBank mRNA | BC110013.1 | - | 1 - 2 | 279 | 74 | |
BT021175.1 | - | 1 - 2 | 328 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC110013.1, NM_001034259.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
