Assay Details
Gene Symbol: C7H1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034272.1 | NP_001029444.1 | 5 - 6 | 497 | 113 | |
GenBank mRNA | BT021507.1 | - | 5 - 6 | 497 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
