Assay Details
| Gene Symbol: ACAA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034319.2 | NP_001029491.1 | 3 - 4 | 422 | 56 | |
| GenBank mRNA | BC102927.1 | - | 3 - 4 | 423 | 56 | |
| EF576938.1 | - | 3 - 4 | 407 | 56 | ||
More Information
Set Membership: |
|

ACAA1
Genomic Map