Assay Details
| Gene Symbol: PSMD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034423.2 | NP_001029595.1 | 6 - 7 | 640 | 62 | |
| GenBank mRNA | BC103437.1 | - | 6 - 7 | 641 | 62 | |
| BT029898.1 | - | 6 - 7 | 607 | 62 | ||
More Information
Set Membership: |
|

PSMD7
Genomic Map