Assay Details
| Gene Symbol: ACTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034502.1 | NP_001029674.1 | 2 - 3 | 190 | 69 | |
| GenBank mRNA | BC102699.1 | - | 2 - 3 | 190 | 69 | |
| BC140575.1 | - | 2 - 3 | 858 | 69 | ||
More Information
Set Membership: |
|

ACTA2
Genomic Map