Assay Details
| Gene Symbol: TMEM9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034507.2 | NP_001029679.1 | 5 - 6 | 638 | 66 | |
| GenBank mRNA | BC102506.1 | - | 5 - 6 | 639 | 66 | |
More Information
Set Membership: |
|

TMEM9B
Genomic Map