Assay Details
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034568.2 | NP_001029740.1 | 1 - 2 | 171 | 99 | |
GenBank mRNA | BC103257.1 | - | 1 - 2 | 172 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103257.1, NM_001034568.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
