Assay Details
| Gene Symbol: CD74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034735.1 | NP_001029907.1 | 4 - 5 | 528 | 60 | |
| GenBank mRNA | BC102388.1 | - | 4 - 5 | 449 | 60 | |
| BT021489.1 | - | 4 - 5 | 528 | 60 | ||
More Information
Set Membership: |
|

CD74
Genomic Map