Assay Details
Gene Symbol: ARPC5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034737.1 | NP_001029909.1 | 3 - 4 | 483 | 143 | |
GenBank mRNA | BC120185.1 | - | 3 - 4 | 497 | 143 | |
BT021057.1 | - | 3 - 4 | 483 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
