Assay Details
| Gene Symbol: MED22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034742.1 | NP_001029914.1 | 1 - 2 | 54 | 94 | |
| GenBank mRNA | BC148153.1 | - | 1 - 2 | 81 | 94 | |
| BT020918.1 | - | 1 - 2 | 54 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC148153.1, NM_001034742.1 transcripts.
Set Membership: |
|

MED22
Genomic Map