Assay Details
Gene Symbol: CARD19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034753.1 | NP_001029925.1 | 3 - 4 | 530 | 66 | |
GenBank mRNA | BT021706.1 | - | 3 - 4 | 530 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
