Assay Details
| Gene Symbol: PPCDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034773.1 | NP_001029945.1 | 2 - 3 | 456 | 75 | |
| GenBank mRNA | BC113213.1 | - | 2 - 3 | 346 | 75 | |
| BT021679.1 | - | 2 - 3 | 456 | 75 | ||
More Information
Set Membership: |
|

PPCDC
Genomic Map