Assay Details
| Gene Symbol: ADAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034780.1 | NP_001029952.1 | 2 - 3 | 211 | 113 | |
| GenBank mRNA | BT020923.1 | - | 2 - 3 | 211 | 113 | |
More Information
Set Membership: |
|

ADAT2
Genomic Map