Assay Details
| Gene Symbol: ADAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034780.1 | NP_001029952.1 | 4 - 5 | 469 | 65 | |
| GenBank mRNA | BT020923.1 | - | 4 - 5 | 469 | 65 | |
More Information
Set Membership: |
|

ADAT2
Genomic Map