Assay Details
Gene Symbol: SKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034781.2 | NP_001029953.1 | 2 - 3 | 202 | 75 | |
GenBank mRNA | BC102435.1 | - | 2 - 3 | 205 | 75 | |
BT029901.1 | - | 2 - 3 | 180 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
