Assay Details
| Gene Symbol: CENPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034782.1 | NP_001029954.1 | 2 - 3 | 563 | 81 | |
| GenBank mRNA | BC126617.1 | - | 2 - 3 | 617 | 81 | |
| BT020751.1 | - | 2 - 3 | 563 | 81 | ||
More Information
Set Membership: |
|

CENPL
Genomic Map