Assay Details
| Gene Symbol: RPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035014.2 | NP_001030186.1 | 2 - 3 | 126 | 88 | |
| GenBank mRNA | BC102556.1 | - | 2 - 3 | 127 | 88 | |
| BC148014.1 | - | 2 - 3 | 129 | 88 | ||
More Information
Set Membership: |
|

RPL23
Genomic Map