Assay Details
| Gene Symbol: RPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035014.2 | NP_001030186.1 | 4 - 5 | 370 | 123 | |
| GenBank mRNA | BC102556.1 | - | 4 - 5 | 371 | 123 | |
| BC148014.1 | - | 4 - 5 | 373 | 123 | ||
| DQ347609.1 | - | 2 - 3 | 193 | 123 | ||
More Information
Set Membership: |
|

RPL23
Genomic Map