Assay Details
| Gene Symbol: SUCLG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035082.1 | NP_001030254.1 | 5 - 6 | 588 | 60 | |
| GenBank mRNA | BT021529.1 | - | 5 - 6 | 588 | 60 | |
More Information
Set Membership: |
|

SUCLG1
Genomic Map