Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035084.2 | NP_001030256.1 | 2 - 3 | 395 | 53 | |
GenBank mRNA | BC103097.1 | - | 2 - 3 | 397 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
