Assay Details
| Gene Symbol: GLRX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035101.1 | NP_001030273.1 | 6 - 7 | 717 | 96 | |
| GenBank mRNA | BC133428.1 | - | 6 - 7 | 730 | 96 | |
| BT021690.1 | - | 6 - 7 | 717 | 96 | ||
More Information
Set Membership: |
|

GLRX3
Genomic Map