Assay Details
| Gene Symbol: DFFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035109.1 | NP_001030281.1 | 5 - 6 | 803 | 66 | |
| GenBank mRNA | BC149123.1 | - | 6 - 7 | 855 | 66 | |
| BT021807.1 | - | 5 - 6 | 803 | 66 | ||
More Information
Set Membership: |
|

DFFB
Genomic Map