Assay Details
| Gene Symbol: SERPINB8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035287.1 | NP_001030364.1 | 3 - 4 | 368 | 128 | |
| GenBank mRNA | BC118173.1 | - | 3 - 4 | 687 | 128 | |
| BT021159.1 | - | 3 - 4 | 368 | 128 | ||
More Information
Set Membership: |
|

SERPINB8
Genomic Map