Assay Details
Gene Symbol: SLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035294.1 | NP_001030371.1 | 3 - 4 | 521 | 51 | |
GenBank mRNA | BC126586.1 | - | 3 - 4 | 555 | 51 | |
BT021197.1 | - | 3 - 4 | 521 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
