Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035324.2 | NP_001030401.1 | 3 - 4 | 267 | 73 | |
GenBank mRNA | BC102484.1 | - | 3 - 4 | 268 | 73 | |
BT025376.1 | - | 3 - 4 | 235 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
