Assay Details
Gene Symbol: KLHDC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035370.2 | NP_001030447.2 | 3 - 4 | 730 | 84 | |
GenBank mRNA | BT020815.1 | - | 3 - 4 | 730 | 84 | |
BT026296.1 | - | 3 - 4 | 720 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
