Assay Details
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035403.1 | NP_001030480.1 | 1 - 2 | 220 | 99 | |
GenBank mRNA | BT021496.1 | - | 1 - 2 | 220 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001035403.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



