Assay Details
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035403.1 | NP_001030480.1 | 2 - 3 | 882 | 144 | |
GenBank mRNA | BC153222.1 | - | 2 - 3 | 809 | 144 | |
BT021496.1 | - | 2 - 3 | 882 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Molecular Function -



