Assay Details
| Gene Symbol: RP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035403.1 | NP_001030480.1 | 3 - 4 | 1005 | 66 | |
| GenBank mRNA | BC153222.1 | - | 3 - 4 | 932 | 66 | |
| BT021496.1 | - | 3 - 4 | 1005 | 66 | ||
More Information
Set Membership: |
|

RP2
Genomic Map