Assay Details
| Gene Symbol: MRPL30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035443.1 | NP_001030520.1 | 3 - 4 | 271 | 77 | |
| GenBank mRNA | BC112734.1 | - | 3 - 4 | 328 | 77 | |
| BT021492.1 | - | 3 - 4 | 271 | 77 | ||
More Information
Set Membership: |
|

MRPL30
Genomic Map